accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68208-18-4

IUPAC Name: (3R)-3-azaniumyl-3-(4-methylphenyl)propanoate
Synonyms: (3R)-3-ammonio-3-(4-methylphenyl)propanoate
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: XPDAKEOBPKFUAH-SECBINFHSA-N
SMILES: CC1=CC=C(C=C1)[C@H]([NH3+])CC([O-])=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)