accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68217-29-8

IUPAC Name: 5-amino-1,4-dioxo-1,4-dihydronaphthalene-2,3-dicarbonitrile
Synonyms: 5-amino-1,4-dioxonaphthalene-2,3-dicarbonitrile
Molecular Weight (g/mol): 223.19
Molecular Formula: C12H5N3O2
InChi Key: BNBGDHHYNYTXFH-UHFFFAOYSA-N
SMILES: NC1=CC=CC2=C1C(=O)C(C#N)=C(C#N)C2=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results