Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2,3-dicyano-1,4-naphthoquinone 97.0+%, TCI America™
SDP

Catalog No. A13601G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A13601G Supplier TCI America Supplier No. A13601G

Chemical Identifiers

CAS 68217-29-8
Molecular Formula C12H5N3O2
Molecular Weight (g/mol) 223.191
MDL Number MFCD00671485
InChI Key BNBGDHHYNYTXFH-UHFFFAOYSA-N
PubChem CID 613915
IUPAC Name 5-amino-1,4-dioxonaphthalene-2,3-dicarbonitrile
SMILES C1=CC2=C(C(=C1)N)C(=O)C(=C(C2=O)C#N)C#N

Specifications

Color Black-Blue
Quantity 1 g
Formula Weight 223.19
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2,3-dicyano-1,4-naphthoquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.