accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68223-03-0

IUPAC Name: 2-{[(benzyloxy)carbonyl](methyl)amino}propanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl](methyl)amino}propanoic acid
Molecular Weight (g/mol): 237.26
Molecular Formula: C12H15NO4
InChi Key: QGEQKVZQPWSOTI-UHFFFAOYNA-N
SMILES: CC(N(C)C(=O)OCC1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)