accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68321-41-5

IUPAC Name: 4-(2-chloroacetyl)-3,4-dihydro-2H-1,4-benzothiazin-3-one
Synonyms: 4-(2-chloroacetyl)-2H-1,4-benzothiazin-3-one
Molecular Weight (g/mol): 241.69
Molecular Formula: C10H8ClNO2S
InChi Key: CYIZUSMRAHFMOI-UHFFFAOYSA-N
SMILES: ClCC(=O)N1C(=O)CSC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results