accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHN

CAS RN 68339-45-7

IUPAC Name: N-[(pyridin-2-yl)methyl]cyclohexanamine
Synonyms: N-(pyridin-2-ylmethyl)cyclohexanamine
Molecular Weight (g/mol): 190.29
Molecular Formula: C12H18N2
InChi Key: KUBDCHSZNMFKFV-UHFFFAOYSA-N
SMILES: C(NC1CCCCC1)C1=CC=CC=N1
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results