Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Cyclohexylaminomethyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. C16855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C16855G Supplier TCI America Supplier No. C16855G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic organic building block for research.

Chemical Identifiers

CAS 68339-45-7
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD01135534
InChI Key KUBDCHSZNMFKFV-UHFFFAOYSA-N
Synonym N-Cyclohexyl-2-picolylamine, N-(2-Pyridylmethyl)cyclohexylamine
PubChem CID 791938
IUPAC Name N-[(pyridin-2-yl)methyl]cyclohexanamine
SMILES C(NC1CCCCC1)C1=CC=CC=N1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 190.29
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material 2-(Cyclohexylaminomethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.