accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3ClClOONFO

CAS RN 68359-37-5

IUPAC Name: cyano(4-fluoro-3-phenoxyphenyl)methyl 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
Synonyms: β-cyfluthrin cyfluthrine epa pesticide chemical code 128831 tempo 2 eulan SP cylence cyfoxylate solfac cyfluthine cyfluthrin
Molecular Weight (g/mol): 434.29
Molecular Formula: C22H18Cl2FNO3
InChi Key: QQODLKZGRKWIFG-UHFFFAOYNA-N
SMILES: CC1(C)C(C=C(Cl)Cl)C1C(=O)OC(C#N)C1=CC=C(F)C(OC2=CC=CC=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results