accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3(S)N(R)OSOOHOO

CAS RN 68373-14-8

IUPAC Name: (2S,5R)-3,3-dimethyl-4,4,7-trioxo-4λ⁶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Synonyms: sulbactam sulbactam sodium (substance) sulbactam sodium sulfone, penicillanic acid combactam sulbactam (product) sulbactam preparation sulbactam (substance) penicillanic acid sulfone sodium, sulbactam
Molecular Weight (g/mol): 233.24
Molecular Formula: C8H11NO5S
InChi Key: FKENQMMABCRJMK-RITPCOANSA-N
SMILES: CC1(C)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(O)=O
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)