Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Sulbactam

Catalog No. AC455350250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455350250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC455350250 Supplier Thermo Scientific Chemicals Supplier No. 455350250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68373-14-8
Molecular Formula C8H11NO5S
Molecular Weight (g/mol) 233.24
InChI Key FKENQMMABCRJMK-RITPCOANSA-N
Synonym sulbactam, sulbactam acid, penicillanic acid sulfone, sulbactamum, betamaze, 2s,5r-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylic acid 4,4-dioxide, penicillanic acid 1,1-dioxide, unii-s4tf6i2330, penicillanic acid dioxide, chembl403
PubChem CID 130313
ChEBI CHEBI:9321
IUPAC Name (2S,5R)-3,3-dimethyl-4,4,7-trioxo-4$l^{6}-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
SMILES CC1(C)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(O)=O

Specifications

Melting Point 156.0°C
Infrared Spectrum Authentic
Quantity 25 g
Formula Weight 233.24
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Sulbactam
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 269-878-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.