accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68399-79-1

IUPAC Name: 2-hydroxy-3-[(1-hydroxy-2-methylpropan-2-yl)amino]propane-1-sulfonic acid
Synonyms: 2-hydroxy-3-[(1-hydroxy-2-methylpropan-2-yl)amino]propane-1-sulfonic acid
Molecular Weight (g/mol): 227.28
Molecular Formula: C7H17NO5S
InChi Key: ACERFIHBIWMFOR-UHFFFAOYNA-N
SMILES: CC(C)(CO)NCC(O)CS(O)(=O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)