Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals AMPSO, 98+%

Catalog No. AAJ6237822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6237822 100 g
AAJ6237814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6237822 Supplier Thermo Scientific Chemicals Supplier No. J6237822
Add to Cart

Useful as a buffer | CAS: 68399-79-1 | C7H17NO5S | 227.28 g/mol

It is used as an alkaline transfer buffer used for the transfer of strongly basic proteins from gels to nitrocellulose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an alkaline transfer buffer used for the transfer of strongly basic proteins from gels to nitrocellulose.

Solubility
Soluble in water (333.33 mg/ml, clear to slightly hazy colorless solution).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68399-79-1
Molecular Formula C7H17NO5S
Molecular Weight (g/mol) 227.28
MDL Number MFCD00041777
InChI Key ACERFIHBIWMFOR-UHFFFAOYNA-N
Synonym N-(1,1-Dimethyl-2-hydroxyethyl)-3-amino-2-hydroxypropanesulfonic acid
PubChem CID 100211
IUPAC Name 2-hydroxy-3-[(1-hydroxy-2-methylpropan-2-yl)amino]propane-1-sulfonic acid
SMILES CC(C)(CO)NCC(O)CS(O)(=O)=O

Specifications

Melting Point 215°C to 225°C
Color White
Physical Form Crystals
Quantity 100 g
Beilstein 5929916
Solubility Information Soluble in water (333.33mg/ml,clear to slightly hazy colorless solution).
Formula Weight 227.28
Percent Purity ≥98%
Chemical Name or Material AMPSO
EINECSNumber 269-991-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.