accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68399-80-4

IUPAC Name: 3-[bis(2-hydroxyethyl)amino]-2-hydroxypropane-1-sulfonic acid
Synonyms: 3-[bis(2-hydroxyethyl)amino]-2-hydroxypropane-1-sulfonic acid
Molecular Weight (g/mol): 243.27
Molecular Formula: C7H17NO6S
InChi Key: XCBLFURAFHFFJF-UHFFFAOYNA-N
SMILES: OCCN(CCO)CC(O)CS(O)(=O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)