accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHOFFF

CAS RN 684-07-1

IUPAC Name: 3,3,3-trifluoro-2-hydroxypropanoic acid
Synonyms: 3,3,3-trifluoro-2-hydroxypropanoic acid
Molecular Weight (g/mol): 144.05
Molecular Formula: C3H3F3O3
InChi Key: BVKGUTLIPHZYCX-UHFFFAOYNA-N
SMILES: OC(C(O)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results