Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,3,3-Trifluoro-2-hydroxypropanoic acid

Catalog No. CDS002724 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS002724 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS002724 Supplier Sigma Aldrich Supplier No. CDS002724100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4,4-dimethyl-2-cyclopenten-1-one is an α,α-unsaturated carbonyl compound and was reported to exhibit the higher tumor-specific cytotoxicity against oral human normal and tumor cells
  • 4,4-dimethyl-2-cyclopenten-1-one was identified as one of the constituents in the essential oil of tagetes minuta L by GC-ms

Specifications

CAS 684-07-1
Molecular Formula C3H3F3O3
Linear Formula C3H3F3O3
MDL Number MFCD00051802
Quantity 1 g
Molecular Weight (g/mol) 144.05
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.