accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 684-78-6

IUPAC Name: 2,2,2-trifluoroacetamido 2,2,2-trifluoroacetate
Synonyms: 2,2,2-trifluoroacetamido 2,2,2-trifluoroacetate
Molecular Weight (g/mol): 225.05
Molecular Formula: C4HF6NO3
InChi Key: CYSAMXITKBXZOP-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)NOC(=O)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results