Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,O-Bis(trifluoroacetyl)hydroxylamine 98.0+%, TCI America™
SDP

Catalog No. B11275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11275G Supplier TCI America Supplier No. B11275G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 684-78-6
Molecular Formula C4HF6NO3
Molecular Weight (g/mol) 225.046
MDL Number MFCD00013565
InChI Key CYSAMXITKBXZOP-UHFFFAOYSA-N
PubChem CID 69634
IUPAC Name [(2,2,2-trifluoroacetyl)amino] 2,2,2-trifluoroacetate
SMILES C(=O)(C(F)(F)F)NOC(=O)C(F)(F)F

Specifications

Melting Point 60°C
Color White-Yellow
Quantity 5 g
Formula Weight 225.05
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,O-Bis(trifluoroacetyl)hydroxylamine
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.