accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNHNNNOO(E)(E/Z)

CAS RN 68420-26-8

IUPAC Name: (E)-N-[(4-nitrophenyl)imino]-N'-(phenylamino)ethanimidamide
Synonyms: (E)-N-[(4-nitrophenyl)imino]-N'-(phenylamino)ethanimidamide
Molecular Weight (g/mol): 283.29
Molecular Formula: C14H13N5O2
InChi Key: FHIGZTOTTOEVNT-DNTUAUKYSA-N
SMILES: C\C(=N/NC1=CC=CC=C1)N=NC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results