accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOHONHNNN(S)

CAS RN 684270-46-0

IUPAC Name: (2S)-3-azido-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2S)-3-azido-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 352.35
Molecular Formula: C18H16N4O4
InChi Key: ZITYCUDVCWLHPG-INIZCTEOSA-N
SMILES: OC(=O)[C@H](CN=[N+]=[N-])NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)