Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Allyldiphenylphosphine

Catalog No. 3368742G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-687-42G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 33-687-42G Supplier Sigma Aldrich Supplier No. 3368742G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cocatalyst in palladium catalyzed hydrocarboxylation reactionsCocatalyst in Palladium catalyzed cross-coupling reactionsLigand for hydroformylation catalystsLigand for the rhenium phosphinoborane pendant Lewis acid-assisted reductive coupling reactionsCatalyst precursor for alkene hydroboration

Specifications

CAS 684270-46-0
Molecular Formula C18H16N4O4
Refractive Index n20/D 1.619
Linear Formula C18H16N4O4
MDL Number MFCD11052919
Quantity 250 mg
Synonym Diphenyl-2-propenylphosphine; NSC 180819; NSC 616248
Molecular Weight (g/mol) 352.34 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.