CAS RN 68441-33-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Supelco™ Amberlite™ IR120, Na+ Form
Strongly acidic cation exchange resin for decalicification.