Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Amberlite™ IR120, Na+ Form

Catalog No. 111015172 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5172 1 kg
11-101-5171 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5172 Supplier MilliporeSigma™ Supelco™ Supplier No. 2243591KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Strongly acidic cation exchange resin for decalicification.

Amberlite™ IR120 is a strong basic anion exchange resin which may be used for the recovery of toxic metals. It may be used in column chromatography. It is required for a ion-exchange resin to swell in the solvent for a higher rate of sorption and to exchange ions.
TRUSTED_SUSTAINABILITY

Specifications

Packaging Poly Bottle
Quantity 1 kg
Product Line Amberlite
Capacity (Metric) 2.0 meq/mL by wetted bed volume
MDL Number MFCD00132707
CAS 68441-33-8

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.