accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OON

CAS RN 68453-37-2

IUPAC Name: methyl 4-[(piperidin-1-yl)methyl]benzoate
Synonyms: methyl 4-(piperidin-1-ylmethyl)benzoate
Molecular Weight (g/mol): 233.31
Molecular Formula: C14H19NO2
InChi Key: VSUPXUIIYYFQDG-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(CN2CCCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results