CAS RN 68480-25-1
IUPAC Name:
(2E)-tridec-2-en-1-ol
Synonyms:
(2E)-tridec-2-en-1-ol
Molecular Weight (g/mol):
198.35
Molecular Formula:
C13H26O
InChi Key:
VPYJHNADOJDSGU-VAWYXSNFSA-N
SMILES:
CCCCCCCCCC\C=C\CO
1
–
1
of
1
results
1
trans-2-Tridecen-1-ol 90.0+%, TCI America™
CAS: 68480-25-1 Molecular Formula: C13H26O Molecular Weight (g/mol): 198.35 MDL Number: MFCD00014048 InChI Key: VPYJHNADOJDSGU-VAWYXSNFSA-N PubChem CID: 5364949 IUPAC Name: (2E)-tridec-2-en-1-ol SMILES: CCCCCCCCCC\C=C\CO