Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Tridecen-1-ol 90.0+%, TCI America™
SDP

Catalog No. T150210ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T150210ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T150210ML Supplier TCI America Supplier No. T150210ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 68480-25-1
Molecular Formula C13H26O
Molecular Weight (g/mol) 198.35
MDL Number MFCD00014048
InChI Key VPYJHNADOJDSGU-VAWYXSNFSA-N
PubChem CID 5364949
IUPAC Name (2E)-tridec-2-en-1-ol
SMILES CCCCCCCCCC\C=C\CO

Specifications

Color Yellow
Quantity 10 mL
Formula Weight 198.35
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Tridecen-1-ol
EINECSNumber (2)-0258
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.