accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHOCH3H3CCH3

CAS RN 68489-09-8

IUPAC Name: N-(4-methoxyphenyl)-5-methyl-2-(propan-2-yl)cyclohexane-1-carboxamide
Synonyms: 2-isopropyl-N-(4-methoxyphenyl)-5-methylcyclohexane-1-carboxamide
Molecular Weight (g/mol): 289.42
Molecular Formula: C18H27NO2
InChi Key: HNSGVPAAXJJOPQ-UHFFFAOYNA-N
SMILES: COC1=CC=C(NC(=O)C2CC(C)CCC2C(C)C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)