accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6850-57-3

IUPAC Name: (2-methoxyphenyl)methanaminium
Synonyms: (2-methoxyphenyl)methanaminium
Molecular Weight (g/mol): 138.19
Molecular Formula: C8H12NO
InChi Key: PXJACNDVRNAFHD-UHFFFAOYSA-O
SMILES: COC1=CC=CC=C1C[NH3+]
Boiling Point 
  • (2)
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)