accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOHNOH3CCH3

CAS RN 6851-81-6

IUPAC Name: 1,5,5-trimethylimidazolidine-2,4-dione
Synonyms: 1,5,5-trimethylimidazolidine-2,4-dione
Molecular Weight (g/mol): 142.16
Molecular Formula: C6H10N2O2
InChi Key: ZNYIPTYJBRGSSL-UHFFFAOYSA-N
SMILES: CN1C(=O)NC(=O)C1(C)C
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)