Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5,5-Trimethylhydantoin 98.0+%, TCI America™
SDP

Catalog No. T17395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17395G 5 g
T173925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17395G Supplier TCI America Supplier No. T17395G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6851-81-6
Molecular Formula C6H10N2O2
Molecular Weight (g/mol) 142.158
MDL Number MFCD00040439
InChI Key ZNYIPTYJBRGSSL-UHFFFAOYSA-N
Synonym 1,5,5-trimethylhydantoin, 3,4,4-trimethyl-2,5-dioxoimidazolidine, 1,5,5-trimethyl-imidazolidine-2,4-dione, 1,5,5-trimethyl-1,3-diazolidine-2,4-dione, hydantoin, 1,5,5-trimethyl, 2,4-imidazolidinedione, 1,5,5-trimethyl, 1,5,5 trimethylhydantoin, acmc-209o3u, 1,5,5-trimethyl-hydantoin, 1,5,5-trimethyl hydantoin
PubChem CID 81295
IUPAC Name 1,5,5-trimethylimidazolidine-2,4-dione
SMILES CC1(C(=O)NC(=O)N1C)C

Specifications

Melting Point 164°C
Color White
Quantity 5 g
Formula Weight 142.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,5,5-Trimethylhydantoin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.