accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68539-16-2

CH3OOOOOH3CH3COOHCH3H2O

CAS RN 68539-16-2

IUPAC Name: 4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[6.4.0.0²,⁶]dodecane-6-carboxylic acid hydrate
Synonyms: 4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[6.4.0.0²,⁶]dodecane-6-carboxylic acid hydrate
Molecular Weight (g/mol): 292.28
Molecular Formula: C12H20O8
InChi Key: ZFQRGFMVXLSLKZ-UHFFFAOYNA-N
SMILES: O.CC1(C)OC2C3OC(C)(C)OCC3OC2(O1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)