Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(-)-2-Amino-3-Methylbutane

Catalog No. 182415GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-241-5GF 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 18-241-5GF Supplier Sigma Aldrich Supplier No. 182415GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-(-)-2-Amino-3-Methylbutane

Specifications

CAS 68539-16-2
Melting Point 186°C
Molecular Formula C9H11NO3 -+ xH2O
Linear Formula C6H5CH(OH)CH(NH2)CO2H-+xH2O
MDL Number MFCD00149295
Quantity 25 g
Synonym (R)-(-)-3-Methyl-2-butylamine
Molecular Weight (g/mol) 181.19 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.