accessibility menu, dialog, popup

UserName

Viewing profile as user:
NIFF

CAS RN 685517-67-3

IUPAC Name: 2,6-difluoro-3-iodopyridine
Synonyms: 2,6-difluoro-3-iodopyridine
Molecular Weight (g/mol): 240.98
Molecular Formula: C5H2F2IN
InChi Key: UTWLONSABCGCBR-UHFFFAOYSA-N
SMILES: FC1=NC(F)=C(I)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results