Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluoro-3-iodopyridine 98.0+%, TCI America™
SDP

Catalog No. D51415G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51415G 5 g
D51411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51415G Supplier TCI America Supplier No. D51415G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 685517-67-3 | C5H2F2IN | 240.979 g/mol

Chemical Identifiers

CAS 685517-67-3
Molecular Formula C5H2F2IN
Molecular Weight (g/mol) 240.979
MDL Number MFCD09743595
InChI Key UTWLONSABCGCBR-UHFFFAOYSA-N
PubChem CID 19768433
IUPAC Name 2,6-difluoro-3-iodopyridine
SMILES C1=CC(=NC(=C1I)F)F

Specifications

Melting Point 42°C
Color White-Yellow
Quantity 5 g
Formula Weight 240.98
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Difluoro-3-iodopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.