accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3CH3

CAS RN 68555-60-2

IUPAC Name: 3-methylbutyl oct-2-ynoate
Synonyms: 3-methylbutyl oct-2-ynoate
Molecular Weight (g/mol): 210.32
Molecular Formula: C13H22O2
InChi Key: OTJSOSMJTUKXJR-UHFFFAOYSA-N
SMILES: CCCCCC#CC(=O)OCCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results