Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl 2-Octynoate, TCI America™
SDP

Catalog No. O025425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O025425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O025425ML Supplier TCI America Supplier No. O025425ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 68555-60-2
Molecular Formula C13H22O2
Molecular Weight (g/mol) 210.317
MDL Number MFCD00036670
InChI Key OTJSOSMJTUKXJR-UHFFFAOYSA-N
Synonym 2-Octynoic Acid Isoamyl Ester, Isopentyl 2-Octynoate, 2-Octynoic Acid Isopentyl Ester
PubChem CID 110542
IUPAC Name 3-methylbutyl oct-2-ynoate
SMILES CCCCCC#CC(=O)OCCC(C)C

Specifications

Color Colorless
Boiling Point 149°C
Quantity 25 mL
Formula Weight 210.32
Physical Form Liquid
Chemical Name or Material Isoamyl 2-Octynoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.