accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3OO

CAS RN 68555-63-5

IUPAC Name: 2-methyl-4-oxo-4H-pyran-3-yl propanoate
Synonyms: 2-methyl-4-oxopyran-3-yl propanoate
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: NBKYPRRBTKRGLE-UHFFFAOYSA-N
SMILES: CCC(=O)OC1=C(C)OC=CC1=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)