Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maltol Propionate 98.0+%, TCI America™
SDP

Catalog No. M067925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M067925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M067925G Supplier TCI America Supplier No. M067925G

Chemical Identifiers

CAS 68555-63-5
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00036605
InChI Key NBKYPRRBTKRGLE-UHFFFAOYSA-N
Synonym 3-Hydroxy-2-methyl-4-pyrone Propionate, 2-Methyl-4-oxo-4H-pyran-3-yl Propionate
PubChem CID 110543
IUPAC Name 2-methyl-4-oxo-4H-pyran-3-yl propanoate
SMILES CCC(=O)OC1=C(C)OC=CC1=O

Specifications

Melting Point 49°C
Color White-Yellow
Quantity 25 g
Formula Weight 182.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Maltol Propionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.