accessibility menu, dialog, popup

UserName

Viewing profile as user:
OFFFOSOOFFF

CAS RN 68602-57-3

IUPAC Name: trifluoromethanesulfonyl 2,2,2-trifluoroacetate
Synonyms: trifluoromethanesulfonyl 2,2,2-trifluoroacetate
Molecular Weight (g/mol): 246.08
Molecular Formula: C3F6O4S
InChi Key: FWJGTOABGBFQNT-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)OS(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)