accessibility menu, dialog, popup

UserName

Viewing profile as user:
BaH3COOH3COO

CAS RN 6865-35-6

IUPAC Name: barium(2+) dioctadecanoate
Synonyms: barium(2+) bis(n-octadecanoate)
Molecular Weight (g/mol): 704.28
Molecular Formula: C36H70BaO4
InChi Key: AGXUVMPSUKZYDT-UHFFFAOYSA-L
SMILES: [Ba++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results