Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Stearate, TCI America™
SDP

Catalog No. S023525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S023525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S023525G Supplier TCI America Supplier No. S023525G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6865-35-6
Molecular Formula C36H70BaO4
Molecular Weight (g/mol) 704.279
MDL Number MFCD00050711
InChI Key AGXUVMPSUKZYDT-UHFFFAOYSA-L
Synonym Stearic Acid Barium Salt
PubChem CID 197077
IUPAC Name barium(2+);octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Ba+2]

Specifications

Melting Point 120°C
Color White
Quantity 25 g
Formula Weight 704.28
Physical Form Crystalline Powder
Chemical Name or Material Barium Stearate
EINECSNumber (2)-0611
RTECSNumber WI2840000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.