accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68698-58-8

IUPAC Name: (3E)-hex-3-en-1-yl oct-2-ynoate
Synonyms: (3E)-hex-3-en-1-yl oct-2-ynoate
Molecular Weight (g/mol): 222.33
Molecular Formula: C14H22O2
InChi Key: ANJQMBWSXVCLTG-SOFGYWHQSA-N
SMILES: CCCCCC#CC(=O)OCC\C=C\CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results