Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-3-Hexen-1-yl 2-Octynoate, TCI America™
SDP

Catalog No. O025525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O025525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O025525ML Supplier TCI America Supplier No. O025525ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 68698-58-8
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.328
MDL Number MFCD00036654
InChI Key ANJQMBWSXVCLTG-SOFGYWHQSA-N
Synonym 2-Octynoic Acid cis-3-Hexen-1-yl Ester
PubChem CID 6437654
IUPAC Name [(E)-hex-3-enyl] oct-2-ynoate
SMILES CCCCCC#CC(=O)OCCC=CCC

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 222.33
Physical Form Liquid
Chemical Name or Material cis-3-Hexen-1-yl 2-Octynoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.