accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6872-17-9

IUPAC Name: 5-chloro-1,3,3-trimethyl-2-methylidene-2,3-dihydro-1H-indole
Synonyms: 5-chloro-1,3,3-trimethyl-2-methylideneindole
Molecular Weight (g/mol): 207.7
Molecular Formula: C12H14ClN
InChi Key: VDMXGJJMPKAYQP-UHFFFAOYSA-N
SMILES: CN1C2=CC=C(Cl)C=C2C(C)(C)C1=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results