Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-1,3,3-trimethyl-2-methyleneindoline 95.0+%, TCI America™
SDP

Catalog No. C117725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C117725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C117725G Supplier TCI America Supplier No. C117725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6872-17-9
Molecular Formula C12H14ClN
Molecular Weight (g/mol) 207.701
MDL Number MFCD00005814
InChI Key VDMXGJJMPKAYQP-UHFFFAOYSA-N
Synonym 5-Chloro-2,3-dihydro-1,3,3-trimethyl-2-methylene-1H-indole
PubChem CID 81308
IUPAC Name 5-chloro-1,3,3-trimethyl-2-methylideneindole
SMILES CC1(C(=C)N(C2=C1C=C(C=C2)Cl)C)C

Specifications

Boiling Point 139°C
Quantity 25 g
Formula Weight 207.70
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Chloro-1,3,3-trimethyl-2-methyleneindoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.