accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68774-77-6

IUPAC Name: 8-chloro-[1,2,4]triazolo[4,3-a]pyrazine
Synonyms: 8-chloro-[1,2,4]triazolo[4,3-a]pyrazine
Molecular Weight (g/mol): 154.56
Molecular Formula: C5H3ClN4
InChi Key: VKZJRJVVFDDJKP-UHFFFAOYSA-N
SMILES: ClC1=NC=CN2C=NN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results