Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Chloro-1,2,4-triazolo[4,3-a]pyrazine, 97%, Thermo Scientific™

Catalog No. AAH6334306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6334306 5 g
AAH6334303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6334306 Supplier Thermo Scientific Chemicals Supplier No. H6334306

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68774-77-6
Molecular Formula C5H3ClN4
Molecular Weight (g/mol) 154.557
MDL Number MFCD08272086
InChI Key VKZJRJVVFDDJKP-UHFFFAOYSA-N
PubChem CID 12582612
IUPAC Name 8-chloro-[1,2,4]triazolo[4,3-a]pyrazine
SMILES C1=CN2C=NN=C2C(=N1)Cl

Specifications

Melting Point 215°C to 220°C
Color Brown
Quantity 5 g
Synonym 8-chloro-1,2,4 triazolo 4,3-a pyrazine, 8-chloro 1,2,4 triazolo 4,3-a pyrazine, 8-chlorotriazolo 4,3-a pyrazine, 1,2,4-triazolo 4,3-a pyrazine, 8-chloro, 8-chloro-1,2,4triazolo 4,3-a pyrazine, 1,2,4-triazolo 4,3-a pyrazine,8-chloro, 8-chloro-1,2,4-triazolo 4,3-a pyrazine
Formula Weight 154.56
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 8-Chloro-1,2,4-triazolo[4,3-a]pyrazine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.