CAS RN 68790-38-5
IUPAC Name:
2-{[(tert-butoxy)carbonyl]amino}benzoic acid
Synonyms:
2-[(tert-butoxycarbonyl)amino]benzoic acid
Molecular Weight (g/mol):
237.26
Molecular Formula:
C12H15NO4
InChi Key:
BYGHHEDJDSLEKK-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NC1=CC=CC=C1C(O)=O
1
–
1
of
1
results
1
Sigma Aldrich Fmoc Isothiocyanate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.