Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Fmoc Isothiocyanate

Catalog No. 109195G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-919-5G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 10-919-5G Supplier Sigma Aldrich Supplier No. 109195G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Fmoc isothiocyanate can be used:As a starting material in the preparation of biologically relevant pharmacophores named N-aryl-N'sa-carboalkoxy guanidines.In one of the intermediate steps for the synthesis of N-aryl-N-thiazolyl derivatives.To synthesize cyclic isothiourea derivatives as potent neuropeptide Y (NPY) Y1receptor antagonists.To prepare 2-aminothiazoles, aminobenz-imidazole conjugated thiazoles, and thiazole derived cyclopeptides.Reagent for the synthesis of substituted thioureas from amines

Specifications

CAS 68790-38-5
Melting Point 153°C to 156°C
Molecular Formula C12H15NO4
Linear Formula (CH3)3CO2CNHC6H4CO2H
MDL Number MFCD00151862
Quantity 10 g
Synonym 9-Fluorenylmethoxycarbonyl isothiocyanate; 9-Fluorenylmethyl isothiocyanatoformate
Molecular Weight (g/mol) 237.25 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.