accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 688-73-3

IUPAC Name: tributylstannyl
Synonyms: tributylstannanyl
Molecular Weight (g/mol): 290.06
Molecular Formula: C12H27Sn
InChi Key: PIILXFBHQILWPS-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)CCCC
Boiling Point 
  • (5)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (8)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (3)
  • (8)
  • (5)
  • (3)
  • (3)
  • (8)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)