Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-n-butyltin hydride, 97%

Catalog No. AAA1329809
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
250 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1329809 10 g
AAA1329814 25 g
AAA1329818 50 g
AAA1329822 100 g
AAA1329830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AAA1329809 Supplier Thermo Scientific Chemicals Supplier No. A1329809
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 688-73-3 | C12H27Sn | 290.058 g/mol

Tri-n-butyltin hydride is used as a radical reagent in reductive cleavage, radical dehalogenation and intramolecular radical cyclization. It also promotes intramolecular cyclization leading to isoxazolo-benzaulene ring system. Further, it acts as a reducing agent for the conversion of alkyl halides to hydrocarbons. In addition to this, it is used in a veterinary anthelmintic for poultry and an intermediate to make other butyltin compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butyltin hydride is used as a radical reagent in reductive cleavage, radical dehalogenation and intramolecular radical cyclization. It also promotes intramolecular cyclization leading to isoxazolo-benzaulene ring system. Further, it acts as a reducing agent for the conversion of alkyl halides to hydrocarbons. In addition to this, it is used in a veterinary anthelmintic for poultry and an intermediate to make other butyltin compounds.

Solubility
Immiscible with water.

Notes
Air and moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 688-73-3
Molecular Formula C12H27Sn
Molecular Weight (g/mol) 290.06
MDL Number MFCD00009416
InChI Key PIILXFBHQILWPS-UHFFFAOYSA-N
Synonym tributylstannanyl, tributlytin, tributylstannic hydride, tributyl tin, tri-n-butylstannane hydride, unii-4xdx163p3d, tin, tri-n-butyl-, hydride, tri-n-butylstannyl radical, stannane, tri-n-butyl-, hydride, tributyl-tin
PubChem CID 3032732
IUPAC Name tributylstannyl
SMILES CCCC[Sn](CCCC)CCCC

Specifications

Melting Point 0°C to 1°C
Density 1.098
Boiling Point 68°C to 74°C (0.3 mmHg)
Flash Point 40°C (104°F)
Odor Odorless
Refractive Index 1.473
Quantity 10 g
UN Number UN2929
Beilstein 3587329
Sensitivity Air sensitive; May form a precipitate
Solubility Information Immiscible with water.
Formula Weight 291.07
Percent Purity 97%
Chemical Name or Material Tri-n-butyltin hydride
Show More Show Less
Hazard Category H301-H312-H315-H317-H319-H360FD-H372
Hazard Statement GHS H Statement
H301-H372-H226-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Flammable liquid and vapour.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
EINECSNumber 211-704-4
RTECSNumber WH8675000
TSCA Yes
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.